1-Amino-2-(isopropylsulfonyl)benzene 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Isopropylsulphonyl)aniline, 2-Aminophenyl Isopropyl sulphone

A372187-1G A372187-5G A372187-100G A372187-10G A372187-250MG A372187-25G
103826-498EA 7 USD
103826-498 103837-516 76840-766 76840-768 77195-492 77195-494 77195-496 77195-498
1-Amino-2-(isopropylsulfonyl)benzene 98%
1-Amino-2-(isopropylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂S
MW: 199.2700 g/mol
Melting Pt: 83…84 °C
Flash Pt: 184.3 °C
Storage Temperature: Ambient
MDL Number: MFCD08445603
CAS Number: 76697-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR