1-Amino-1-cyclopropanecarbonitrile hydrochloride 97%

Supplier: Ambeed
Danger

A100890-1G A100890-5G A100890-25G A100890-250MG
103795-678EA 13 USD
103795-678 103821-368 77296-888 77296-890 77570-078 77880-824
1-Amino-1-cyclopropanecarbonitrile hydrochloride 97%
1-Amino-1-cyclopropanecarbonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₄H₇ClN₂
Melting Pt: 214…220 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114063
CAS Number: 127946-77-4
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR