1AH-dibenzo[b,f]oxireno[2,3-d]azepine-6(10bH)-carboxamide 97%

Supplier: Ambeed
Warning

Synonyms: Carbamazepine 10,11-epoxide

A267353-1G A267353-250MG A267353-100MG
103799-544EA 68 USD
103799-544 103812-846 77304-972 77601-148 77668-156
1AH-dibenzo[b,f]oxireno[2,3-d]azepine-6(10bH)-carboxamide 97%
1AH-dibenzo[b,f]oxireno[2,3-d]azepine-6(10bH)-carboxamide

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₂O₂
MW: 252.26 g/mol
Boiling Pt: 390.2 °C (760 mm Hg)
Melting Pt: 204…206 °C
Flash Pt: 189.8 °C
Storage Temperature: Freezer
MDL Number: MFCD00467137
CAS Number: 36507-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR