18-(tert-Butoxy)-18-oxooctadecanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl Hydrogen Octadecanedioate, Mono-tert-butyl Octadecanedioate, tBu ester-C16-acid, Octadecanedioic acid Mono-tert-butyl ester

A107224-100G A107224-100MG A107224-250MG A107224-1G
103797-154EA 10 USD
103797-154 103822-920 76717-898 76717-900 76717-902 77283-558
18-(tert-Butoxy)-18-oxooctadecanoic acid 97%
18-(tert-Butoxy)-18-oxooctadecanoic acid

Precaución: for Research Use Only

Formula: C₂₂H₄₂O₄
MW: 370.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09991735
CAS Number: 843666-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR