tert-Butyl Hydrogen Hexadecanedioate 98%

Supplier: Ambeed
Warning

Synonyms: 16-(tert-Butoxy)-16-oxohexadecanoic acid, Hexadecanedioic acid Mono-tert-butyl ester, Boc-C14-COOH

A194358-10G A194358-250MG A194358-1G A194358-25G A194358-100G
103816-644EA 9 USD
103816-644 103847-256 77151-972 77151-974 77302-412 77302-414 77900-501
tert-Butyl Hydrogen Hexadecanedioate 98%
tert-Butyl Hydrogen Hexadecanedioate

Precaución: for Research Use Only

Formula: C₂₀H₃₈O₄
Storage Temperature: Ambient
MDL Number: MFCD23136039
CAS Number: 843666-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR