14H-Anthra[2,1,9-mna]thioxanthen-14-one 95%

Supplier: Ambeed
Warning

A223578-100G A223578-10G A223578-250MG A223578-25G A223578-1G A223578-5G
103819-044EA 7 USD
103819-044 103849-924 76722-030 76722-032 76722-034 76722-036 77088-544 77088-546
14H-Anthra[2,1,9-mna]thioxanthen-14-one 95%
14H-Anthra[2,1,9-mna]thioxanthen-14-one

Precaución: for Research Use Only

Formula: C₂₃H₁₂OS
MW: 336.4 g/mol
Boiling Pt: 607.8 °C (760 mm Hg)
Melting Pt: 306…310 °C
Storage Temperature: Ambient
MDL Number: MFCD00800439
CAS Number: 16294-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR