tert-Butyl Hydrogen Dodecanedioate 97%

Supplier: Ambeed
Warning

Synonyms: 12-(tert-Butoxy)-12-oxododecanoic acid, Dodecanedioic acid Mono-tert-butyl ester

A236332-100MG A236332-10G A236332-250MG A236332-1G A236332-25G A236332-5G
103811-080EA 28 USD
103811-080 103850-852 77153-006 77153-008 77153-010 77304-034 77304-036 77304-038
tert-Butyl Hydrogen Dodecanedioate 97%
tert-Butyl Hydrogen Dodecanedioate

Precaución: for Research Use Only

Formula: C₁₆H₃₀O₄
MW: 286.41 g/mol
Boiling Pt: 389.1±15.0 °C (760 mm Hg)
Flash Pt: 129.6 °C
Storage Temperature: Ambient
MDL Number: MFCD28098251
CAS Number: 234081-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR