11,12-Dihydroindolo[2,3-a]carbazole 96%

Supplier: Ambeed
Warning

A224359-1G A224359-25G A224359-5G A224359-100G
103819-104EA 7 USD
103819-104 103849-994 77088-642 77088-644 77088-646 77192-978
11,12-Dihydroindolo[2,3-a]carbazole 96%
11,12-Dihydroindolo[2,3-a]carbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₂N₂
MW: 256.31 g/mol
Boiling Pt: 569.833 °C (760 mm Hg)
Flash Pt: 269.854 °C
Storage Temperature: Ambient
MDL Number: MFCD07371389
CAS Number: 60511-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR