11-(1-Piperazinyl)dibenzo[b,f][1,4]thiazepine dihydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Norquetiapine 2HCl, Norquetiapine dihydrochloride, 11-(1-Piperazinyl)-dibenzo[b,f][1,4]thiazepine dihydrochloride

A111661-100G A111661-10G A111661-5G A111661-1G
103797-880EA 18 USD
103797-880 103823-700 76995-252 76995-254 77284-156 77310-512
11-(1-Piperazinyl)dibenzo[b,f][1,4]thiazepine dihydrochloride 95%
11-(1-Piperazinyl)dibenzo[b,f][1,4]thiazepine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₁₉Cl₂N₃S
MW: 368.3239 g/mol
Boiling Pt: 513.3 °C (760 mm Hg)
Melting Pt: 180…184 °C
Flash Pt: 264.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08703302
CAS Number: 111974-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR