10-Phenylphenothiazine 98%

Supplier: Ambeed
Warning

A232078-5G A232078-10G A232078-250MG A232078-1G A232078-25G
103810-812EA 52 USD
103810-812 103850-566 77089-486 77237-912 77237-914 77303-928 77615-924
10-Phenylphenothiazine 98%
10-Phenylphenothiazine

Precaución: for Research Use Only

Formula: C₁₈H₁₃NS
Boiling Pt: 411.2 °C (760 mm Hg)
Flash Pt: 202.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00156238
CAS Number: 7152-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR