10-Hydroxydecanoic acid 98%

Supplier: Ambeed
Warning

A114969-1000G A114969-100G A114969-10G A114969-500G A114969-1G A114969-5G A114969-25G
103798-508EA 5 USD
103798-508 103824-350 77189-310 77189-312 77189-314 77189-316 77311-170 77311-172 77566-052
10-Hydroxydecanoic acid 98%
10-Hydroxydecanoic acid

Precaución: For research use only.

Formula: C₁₀H₂₀O₃
Melting Pt: 75…77 °C
Flash Pt: 168.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00010510
CAS Number: 1679-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR