10-Hydroxy-10,11-dihydro-5H-dibenzo[b,f]azepine-5-carboxamide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Licarbazepine
A334944-1G
A334944-250MG
A334944-5G
A334944-100MG
103825-678EA
101
USD
103825-678
103836-648
76839-656
77032-978
77306-480
77668-534
10-Hydroxy-10,11-dihydro-5H-dibenzo[b,f]azepine-5-carboxamide 97%
10-Hydroxy-10,11-dihydro-5H-dibenzo[b,f]azepine-5-carboxamide
Precaución: for Research Use Only
Formula:
C₁₅H₁₄N₂O₂ MW: 254.28 g/mol Boiling Pt: 431.3 °C (760 mm Hg) Melting Pt: 186…189 °C Flash Pt: 214.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00871817 CAS Number: 29331-92-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...