10-Bromo-1-decanol 96%

Supplier: Ambeed
Warning

Synonyms: 10-Bromodecanol

A113573-1G A113573-5G A113573-100G A113573-25G A113573-10G
103798-252EA 6 USD
103798-252 103824-092 77310-906 77310-908 77549-206 77549-208 77592-206
10-Bromo-1-decanol 96%
10-Bromo-1-decanol

Precaución: for Research Use Only

Formula: HO(CH₂)₉CH₂Br
MW: 237.18 g/mol
Boiling Pt: 288.6 °C (760 mm Hg)
Flash Pt: 118.3 °C
Storage Temperature: Freezer
MDL Number: MFCD00041681
CAS Number: 53463-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR