10-Bromodecanoic acid 95%

Supplier: Ambeed
Warning

A249091-100G A249091-10G A249091-1G A249091-500G A249091-25G A249091-5G
103851-692EA 9 USD
103851-692 76976-640 76976-642 76976-644 76976-646 77091-034 77091-036
10-Bromodecanoic acid 95%
10-Bromodecanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₉BrO₂
Boiling Pt: 339.2 °C (760 mm Hg)
Melting Pt: 38…41 °C
Flash Pt: 158.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00014388
CAS Number: 50530-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR