10-Bromodec-1-ene 98%

Supplier: Ambeed
Warning

A145010-100G A145010-10G A145010-25G A145010-1G A145010-5G
103803-480EA 6 USD
103803-480 103841-558 77190-570 77190-572 77299-432 77316-828 77316-830
10-Bromodec-1-ene 98%
10-Bromodec-1-ene

Precaución: for Research Use Only

Formula: C₁₀H₁₉Br
Flash Pt: 96.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00078660
CAS Number: 62871-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR