10,10-Dimethyl-9,10-dihydroanthracen-9-one 98%
Supplier: Ambeed
|
Warning
|
A196728-100G
A196728-10G
A196728-25G
A196728-5G
A196728-1G
103816-862EA
6
USD
103816-862
103847-476
76975-092
76975-094
77085-446
77085-448
77302-524
10,10-Dimethyl-9,10-dihydroanthracen-9-one 98%
10,10-Dimethyl-9,10-dihydroanthracen-9-one
Precaución: for Research Use Only
Formula:
C₁₆H₁₄O MW: 222.28 g/mol Boiling Pt: 346.6 °C (760 mm Hg) Melting Pt: 101…106 °C Flash Pt: 149.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00204395 CAS Number: 5447-86-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...