10-(tert-Butoxy)-10-oxodecanoic acid 97%

Supplier: Ambeed
Warning

A104959-100MG A104959-10G A104959-250MG A104959-25G A104959-1G A104959-5G
103796-806EA 32 USD
103796-806 103822-532 77093-700 77093-702 77093-704 77093-706 77309-394 77309-396
10-(tert-Butoxy)-10-oxodecanoic acid 97%
10-(tert-Butoxy)-10-oxodecanoic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₆O₄
MW: 258.35 g/mol
Boiling Pt: 355.9 °C (760 mm Hg)
Flash Pt: 121.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD28505598
CAS Number: 234081-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR