10-(Phosphonooxy)decyl methacrylate 95%

Supplier: Ambeed
Warning

A269465-10G A269465-250MG A269465-1G A269465-25G A269465-5G A269465-100G
103812-966EA 8 USD
103812-966 103852-872 76977-154 76977-156 77092-638 77092-640 77092-642 77571-672
10-(Phosphonooxy)decyl methacrylate 95%
10-(Phosphonooxy)decyl methacrylate

Precaución: for Research Use Only

Formula: C₁₄H₂₇O₆P
Boiling Pt: 450.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD19441164
CAS Number: 85590-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR