1-[3-(Trifluoromethyl)phenyl]propylamine 97%

Supplier: Ambeed
Danger

Synonyms: 1-[3-(Trifluoromethyl)phenyl]propan-1-amine

A100181-100MG A100181-1G
103795-282EA 286 USD
103795-282 77270-138 77282-448
1-[3-(Trifluoromethyl)phenyl]propylamine 97%
1-[3-(Trifluoromethyl)phenyl]propylamine

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₃N
MW: 203.204 g/mol
Boiling Pt: 205.5 °C (760 mm Hg)
Flash Pt: 83.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07784289
CAS Number: 473732-59-1
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR