1,8-Naphthyridine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A185765-250MG A185765-100MG
103815-880EA 821 USD
103815-880 77029-606 77871-923
1,8-Naphthyridine-3-carboxylic acid 95%
1,8-Naphthyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆N₂O₂
Boiling Pt: 366.8±27.0 °C (760 mm Hg)
Melting Pt: 304 °C
Flash Pt: 175.6 °C
Storage Temperature: Ambient
MDL Number: MFCD06254513
CAS Number: 104866-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR