1,8-Dichloroisoquinoline 96%

Supplier: Ambeed
Warning

A101940-100MG A101940-250MG A101940-1G
103796-212EA 143 USD
103796-212 103821-908 76821-264 76821-266 77282-866
1,8-Dichloroisoquinoline 96%
1,8-Dichloroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₅Cl₂N
MW: 198.05 g/mol
Boiling Pt: 327.7±22.0 °C (760 mm Hg)
Flash Pt: 182 °C
Storage Temperature: Ambient
MDL Number: MFCD15526764
CAS Number: 848841-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR