1,8-Dibromooctane 98%
Supplier: Ambeed
|
Warning
|
A109682-1000G
A109682-100G
A109682-10G
A109682-1G
A109682-25G
A109682-500G
A109682-5G
103823-350EA
7
USD
103823-350
77188-990
77188-992
77188-994
77188-996
77188-998
77189-000
77283-878
1,8-Dibromooctane 98%
1,8-Dibromooctane
Precaución: for Research Use Only
Formula:
BrH₂C(CH₂)₆CH₂Br Boiling Pt: 270…272 °C (760 mm Hg) Melting Pt: 12…16 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000277 CAS Number: 4549-32-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...