1,8-Dibromonaphthalene 98%

Supplier: Ambeed
Warning

A410357-1G A410357-25G A410357-5G A410357-100G A410357-10G A410357-250MG
103827-210EA 6 USD
103827-210 103838-256 76841-774 76841-776 76841-778 77115-822 77115-824 77115-826
1,8-Dibromonaphthalene 98%
1,8-Dibromonaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆Br₂
Boiling Pt: 339.1 °C (760 mm Hg)
Melting Pt: 106…108 °C
Flash Pt: 184.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00183574
CAS Number: 17135-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR