1,7'-Dimethyl-2'-propyl-1H,1'H-2,5'-bibenzo[d]imidazole 95%

Supplier: Ambeed
Warning

A384338-25G A384338-5G A384338-100G A384338-10G A384338-500G
103810-256EA 156 USD
103810-256 103837-800 76841-150 76841-152 77003-670 77003-672 77003-674
1,7'-Dimethyl-2'-propyl-1H,1'H-2,5'-bibenzo[d]imidazole 95%
1,7'-Dimethyl-2'-propyl-1H,1'H-2,5'-bibenzo[d]imidazole

Precaución: for Research Use Only

Formula: C₁₉H₂₀N₄
MW: 304.39 g/mol
Boiling Pt: 584.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03840857
CAS Number: 152628-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR