1,6-Naphthyridine-2-carboxylic acid 97%

Supplier: Ambeed

A154332-100MG A154332-250MG A154332-25G
103804-628EA 28 USD
103804-628 103842-814 77259-394 77259-396 77318-524
1,6-Naphthyridine-2-carboxylic acid 97%
1,6-Naphthyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆N₂O₂
MDL Number: MFCD00202908
CAS Number: 197507-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR