1,6-Dioxaspiro[2.5]octane 95%

Supplier: Ambeed
Danger

A182800-250MG A182800-1G
103815-570EA 98 USD
103815-570 103846-128 76974-508 77083-694
1,6-Dioxaspiro[2.5]octane 95%
1,6-Dioxaspiro[2.5]octane

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
Boiling Pt: 158.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01730206
CAS Number: 185-72-8
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR