1,6-Diiodohexane 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilized with Copper chip

A116396-100G A116396-25G A116396-500G A116396-5G
103824-590EA 7 USD
103824-590 77189-402 77189-404 77189-406 77311-400
1,6-Diiodohexane 98% stabilized
1,6-Diiodohexane

Precaución: for Research Use Only

Formula: C₆H₁₂I₂
Melting Pt: 9…10 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00001103
CAS Number: 629-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR