1,6-Dibromopyrene 98%

Supplier: Ambeed
Warning

A201510-1G A201510-5G A201510-100G A201510-10G A201510-500G A201510-25G
103817-304EA 6 USD
103817-304 103847-944 77086-016 77086-018 77192-332 77192-334 77192-336 77302-762
1,6-Dibromopyrene 98%
1,6-Dibromopyrene

Precaución: for Research Use Only

Formula: C₁₆H₈Br₂
Boiling Pt: 469.6 °C (760 mm Hg)
Melting Pt: 221…227 °C
Flash Pt: 277.3 °C
Storage Temperature: Ambient
MDL Number: MFCD09263746
CAS Number: 27973-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR