1,5-Naphthyridine 98%

Supplier: Ambeed
Warning

A163920-100G A163920-10G A163920-250MG A163920-1G A163920-25G A163920-5G
103805-790EA 9 USD
103805-790 103844-054 77191-192 77191-194 77191-196 77320-176 77320-178 77320-180
1,5-Naphthyridine 98%
1,5-Naphthyridine

Precaución: Sealed in dry, at room temperature.

Formula: C₈H₆N₂
MW: 130.15 g/mol
Boiling Pt: 248.9 °C (760 mm Hg)
Melting Pt: 74 °C
Flash Pt: 107.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00039727
CAS Number: 254-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR