1,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2-(1,5-Dimethyl-1H-pyrazol-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1,5-Dimethyl-1H-pyrazole-4-boronic acid Pinacol ester

A434561-1G A434561-25G A434561-5G A434561-250MG
103827-578EA 15 USD
103827-578 103838-644 76842-356 76842-358 76842-360 76979-326
1,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester 95%
1,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester

Precaución: For research use only

Formula: C₁₁H₁₉BN₂O₂
MW: 222.0918 g/mol
Melting Pt: 82…86 °C
Storage Temperature: Freezer
MDL Number: MFCD09864189
CAS Number: 1036991-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR