1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazole-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A142145-100MG A142145-250MG
103803-106EA 85 USD
103803-106 77162-374 77162-376
1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazole-4-carboxylic acid 95%
1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00020785
CAS Number: 83-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR