1,5-Dimethoxynaphthalene 96%

Supplier: Ambeed
Warning

A178329-1G A178329-5G A178329-10G
103807-320EA 10 USD
103807-320 103845-660 77083-078 77083-080 77191-616
1,5-Dimethoxynaphthalene 96%
1,5-Dimethoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₂
MW: 188.23 g/mol
Boiling Pt: 311.2 °C (760 mm Hg)
Melting Pt: 180…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00086779
CAS Number: 10075-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR