1,5-Dibromopentane 98%

Supplier: Ambeed
Warning

A154427-100G A154427-500G A154427-25G A154427-5G A154427-1000G
103808-800EA 46 USD
103808-800 103842-826 77150-578 77150-580 77300-058 77300-060 77318-546
1,5-Dibromopentane 98%
1,5-Dibromopentane

Precaución: for Research Use Only

Formula: BrH₂C(CH₂)₃CH₂Br
MW: 229.94 g/mol
Boiling Pt: 110 °C (760 mm Hg)
Melting Pt: …34 °C
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00000268
CAS Number: 111-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR