1,5-Dibromonaphthalene 96%

Supplier: Ambeed
Warning

A111343-250MG A111343-25G A111343-1G A111343-5G A111343-10G
103797-822EA 21 USD
103797-822 103823-638 77025-784 77025-786 77310-444 77310-446 77599-604
1,5-Dibromonaphthalene 96%
1,5-Dibromonaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆Br₂
MW: 285.9600 g/mol
Boiling Pt: 326.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00089984
CAS Number: 7351-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR