1,5-Dibromo-2,4-dimethylbenzene 95%

Supplier: Ambeed
Warning

Synonyms: 4,6-Dibromo-m-xylene

A205572-100G A205572-10G A205572-500G A205572-1G A205572-25G A205572-5G
103817-642EA 6 USD
103817-642 103848-308 76986-564 76986-566 76986-568 77086-526 77086-528 77086-530
1,5-Dibromo-2,4-dimethylbenzene 95%
1,5-Dibromo-2,4-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈Br₂
MW: 263.96 g/mol
Melting Pt: 71…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00154984
CAS Number: 615-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR