1,5-Dibromo-2,4-difluorobenzene 97%

Supplier: Ambeed
Warning

A206094-100G A206094-10G A206094-25G A206094-5G A206094-1G
103817-686EA 5 USD
103817-686 103848-358 77030-296 77030-298 77086-596 77086-598 77302-982
1,5-Dibromo-2,4-difluorobenzene 97%
1,5-Dibromo-2,4-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂F₂
MW: 271.8849 g/mol
Boiling Pt: 201.3 °C (760 mm Hg)
Flash Pt: 75.588 °C
Storage Temperature: Freezer
MDL Number: MFCD08458122
CAS Number: 28342-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR