1,5,2,4-Dioxadithiane 2,2,4,4-tetraoxide 97%

Supplier: Ambeed
Warning

A201111-1G A201111-5G A201111-100G A201111-10G
103817-270EA 5 USD
103817-270 103847-906 77085-982 77085-984 77567-916 77567-918
1,5,2,4-Dioxadithiane 2,2,4,4-tetraoxide 97%
1,5,2,4-Dioxadithiane 2,2,4,4-tetraoxide

Precaución: For research use only.

Formula: C₂H₄O₆S₂
MW: 188.17 g/mol
Boiling Pt: 624.2 °C (760 mm Hg)
Flash Pt: 331.332 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19440777
CAS Number: 99591-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR