1,4-Phenyldiboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: p-Phenylenebisboronic acid, 1,4-Phenylenediboronic acid, 1,4-Phenyldiboric acid, 1,4-Benzenediboronic acid

A219818-25G A219818-5G A219818-100G A219818-10G A219818-1G
103818-820EA 9 USD
103818-820 103849-702 77088-272 77088-274 77112-712 77112-714 77303-534
1,4-Phenyldiboronic acid 98%
1,4-Phenyldiboronic acid

Precaución: for Research Use Only

Formula: C₆H₈B₂O₄
MW: 165.75 g/mol
Melting Pt: >350 °C
Flash Pt: 207.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00236018
CAS Number: 4612-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR