1,4-Oxazepane hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Homomorpholine hydrochloride

A135532-250MG A135532-25G A135532-5G A135532-1G
103802-172EA 34 USD
103802-172 76983-964 77298-792 77298-794 77315-008
1,4-Oxazepane hydrochloride 98%
1,4-Oxazepane hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClNO
MW: 137.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02683070
CAS Number: 178312-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR