1,4-Oxazepan-5-one 97%

Supplier: Ambeed
Warning

A105056-10G A105056-250MG A105056-1G A105056-5G
103796-830EA 30 USD
103796-830 103822-556 77188-732 77188-734 77283-336 77283-338
1,4-Oxazepan-5-one 97%
1,4-Oxazepan-5-one

Precaución: for Research Use Only

Formula: C₅H₉NO₂
MW: 115.13 g/mol
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD02683091
CAS Number: 10341-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR