1,4-Dioxaspiro[4.5]decan-8-ol 98%

Supplier: Ambeed
Warning

A130001-100G A130001-10G A130001-500G A130001-1G A130001-25G A130001-5G
103801-362EA 12 USD
103801-362 77009-868 77009-870 77009-872 77298-382 77313-782 77313-784
1,4-Dioxaspiro[4.5]decan-8-ol 98%
1,4-Dioxaspiro[4.5]decan-8-ol

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
Boiling Pt: 269 °C (760 mm Hg)
Flash Pt: 116.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00067003
CAS Number: 22428-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR