1,4-Dioxane-2,5-dione 97%

Supplier: Ambeed
Warning

Synonyms: Glycolide, 1,4-Dioxane-2,5-dione

A177937-10G A177937-1G
103807-278EA 7 USD
103807-278 103845-618 76799-580 77301-530
1,4-Dioxane-2,5-dione 97%
1,4-Dioxane-2,5-dione

Precaución: for Research Use Only

Formula: C₄H₄O₄
MW: 116.07 g/mol
Boiling Pt: 322.5 °C (760 mm Hg)
Melting Pt: 81…83 °C
Flash Pt: 181.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00081108
CAS Number: 502-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR