1,4-Dioxan-2-one 98%

Supplier: Ambeed
Warning

Synonyms: p-Dioxanone

A100823-100G A100823-10G A100823-25G A100823-500G A100823-1G A100823-5G
103795-640EA 6 USD
103795-640 103821-336 76796-246 76796-248 76796-250 76796-252 77282-576 77282-578
1,4-Dioxan-2-one 98%
1,4-Dioxan-2-one

Precaución: for Research Use Only

Formula: C₄H₆O₃
MW: 102.09 g/mol
Boiling Pt: 222.9 °C (760 mm Hg)
Flash Pt: 85.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00215855
CAS Number: 3041-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR