1,4-Dimethyl-1H-pyrazole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A391982-100MG A391982-250MG A391982-1G
103826-890EA 92 USD
103826-890 103837-928 76978-950 76978-952 77307-004
1,4-Dimethyl-1H-pyrazole-3-carboxylic acid 97%
1,4-Dimethyl-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MW: 140.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19228348
CAS Number: 89202-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR