1,4-Diiodo-2-nitrobenzene 97%
Supplier: Ambeed
|
Warning
|
A128884-250MG
A128884-5G
103801-194EA
13
USD
103801-194
77189-998
77313-552
1,4-Diiodo-2-nitrobenzene 97%
1,4-Diiodo-2-nitrobenzene
Precaución: for Research Use Only
Formula:
C₆H₃I₂NO₂ MW: 374.9 g/mol Boiling Pt: 354.7 °C (760 mm Hg) Melting Pt: 109…110 °C Flash Pt: 168.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD09030977 CAS Number: 89488-57-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...