1,4-Diiodo-2,5-dimethoxybenzene 98%

Supplier: Ambeed
Warning

A195124-1G A195124-25G A195124-5G A195124-100G A195124-10G A195124-250MG
103816-724EA 9 USD
103816-724 103847-338 77085-288 77085-290 77085-292 77192-118 77192-120 77192-122
1,4-Diiodo-2,5-dimethoxybenzene 98%
1,4-Diiodo-2,5-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈I₂O₂
MW: 389.9569 g/mol
Boiling Pt: 382.9 °C (760 mm Hg)
Melting Pt: 171 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00461369
CAS Number: 51560-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR