1,4-Difluoro-2-iodobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Difluoroiodobenzene

A281663-25G A281663-5G
103853-514EA 13 USD
103853-514 76837-206 76837-208
1,4-Difluoro-2-iodobenzene 98%
1,4-Difluoro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃F₂I
MW: 239.99 g/mol
Boiling Pt: 178…179 °C (760 mm Hg)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD03094151
CAS Number: 2265-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR