1,4-Dichloro-2-butyne 97%

Supplier: Ambeed
Warning

Synonyms: 1,4-Dichlorobut-2-yne

A107366-100G A107366-10G A107366-1G A107366-25G A107366-500G A107366-5G
103822-950EA 9 USD
103822-950 77188-866 77188-868 77188-870 77188-872 77188-874 77309-800
1,4-Dichloro-2-butyne 97%
1,4-Dichloro-2-butyne

Precaución: for Research Use Only

Formula: C₄H₄Cl₂
MW: 122.98 g/mol
Boiling Pt: 165.5±0.0 °C (760 mm Hg)
Flash Pt: 70.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00000981
CAS Number: 821-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR