1,4-Dibromotetrafluorobenzene 98%
Supplier: Ambeed
|
Warning
|
A544148-1G
A544148-100G
A544148-10G
A544148-25G
A544148-5G
103828-652EA
9
USD
103828-652
103839-774
76843-830
77197-442
77197-444
77307-778
77307-780
1,4-Dibromotetrafluorobenzene 98%
1,4-Dibromotetrafluorobenzene
Precaución: for Research Use Only
Formula:
C₆Br₂F₄ Boiling Pt: 191.9 °C (760 mm Hg) Melting Pt: 78…81 °C at N/A mmHg Flash Pt: 69.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000309 CAS Number: 344-03-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...