1,4-Dibromonaphthalene 98%

Supplier: Ambeed
Warning

A104918-10G A104918-500G A104918-1G A104918-5G A104918-25G A104918-100G
103796-798EA 5 USD
103796-798 103822-524 76796-450 76796-452 77309-386 77309-388 77549-000 77570-290
1,4-Dibromonaphthalene 98%
1,4-Dibromonaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆Br₂
MW: 285.97 g/mol
Boiling Pt: 310
Melting Pt: 92
Flash Pt: 181.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00041823
CAS Number: 83-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR